Learn More
Description
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Rhodamine |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free |
| Gene Alias | Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) |
| Host Species | Goat |
| Immunogen | This GFP antibody was developed against a fusion protein corresponding to the full length GFP (246aa) derived from the jellyfish Aequorea victoria. |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
