Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Mouse, DyLight 550, Clone: 1025527, Novus Biologicals™
SDP

Catalog No. NB173941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173941 Supplier Novus Biologicals Supplier No. FAB42402L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GFP Monoclonal specifically detects GFP in Multi-species samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot.

Specifications

Antigen GFP
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1025527
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Mouse
Immunogen E. coli-derived recombinant GFP, aa 2-238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.