Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFPT1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB088985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088985 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088985 Supplier Novus Biologicals Supplier No. NBP299692B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFPT1 Polyclonal specifically detects GFPT1 in Human samples. It is validated for ELISA.

Specifications

Antigen GFPT1
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GFPT1 (AAA58502.1; Gln 332-Glu 699).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.