Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-1/GDNF R alpha-1 Antibody (009), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB039455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039455 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039455 Supplier Novus Biologicals Supplier No. NBP290463C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GFR alpha-1/GDNF R alpha-1 Monoclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GFR alpha-1/GDNF R alpha-1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse GFR alpha-1/GDNF R alpha-1 (AAB86600.1; Met 1-Ser 425).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2674
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.