Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-2/GDNF R alpha-2 Antibody (71515) [Biotin], Novus Biologicals™
SDP

Catalog No. NB377007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377007 Supplier Novus Biologicals Supplier No. FAB6131B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFR alpha-2/GDNF R alpha-2 Monoclonal antibody specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen GFR alpha-2/GDNF R alpha-2
Applications Immunocytochemistry
Classification Monoclonal
Clone 71515
Conjugate Biotin
Dilution Immunocytochemistry
Formulation PBS
Gene Alias GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2, Ser22-Ser441, Accession # O00451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2675
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.