Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-3/GDNF R alpha-3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB083043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB083043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB083043 Supplier Novus Biologicals Supplier No. NBP299078AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFR alpha-3/GDNF R alpha-3 Polyclonal specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for ELISA.

Specifications

Antigen GFR alpha-3/GDNF R alpha-3
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GFR alpha-3/GDNF R alpha-3 (NP_001487.2; Met 1-Trp 382).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2676
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.