Learn More
GFR alpha-4/GDNF R alpha-4/Persephin receptor Antibody (MM0311-3P34), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MM0311-3P34 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor |
| Gene Symbols | GFRA4 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.