Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-4/GDNF R alpha-4/Persephin receptor Antibody (MM0311-3P34), FITC, Novus Biologicals™
SDP

Catalog No. NB360947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360947 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360947 Supplier Novus Biologicals Supplier No. NBP212414F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal antibody specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Human samples. It is validated for Western Blot

Specifications

Antigen GFR alpha-4/GDNF R alpha-4/Persephin receptor
Applications Western Blot
Classification Monoclonal
Clone MM0311-3P34
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor
Host Species Mouse
Immunogen Human recombinant GFR alpha-4/GDNF R alpha-4/Persephin receptor
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64096
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.