Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGA2 Antibody (OTI1D2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27172A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27172A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27172A70 Supplier Novus Biologicals Supplier No. NBP271725AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GGA2 Monoclonal specifically detects GGA2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GGA2
Applications Western Blot
Classification Monoclonal
Clone OTI1D2
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ20966, Gamma-adaptin-related protein 2, golgi associated, gamma adaptin ear containing, ARF binding protein 2, golgi-associated, gamma adaptin ear containing, ARF binding protein 2, Golgi-localized, gamma ear-containing, ARF-binding protein 2, KIAA1080ADP-ribosylation factor-binding protein GGA2, VEAR, VHS domain and ear domain of gamma-adaptin, VHS domain and ear domain-containing protein
Gene Symbols GGA2
Host Species Mouse
Immunogen Full length recombinant protein of human GGA2 (NP_055859) produced in HEK293T cells
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23062
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.