Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGA3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB327833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327833 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327833 Supplier Novus Biologicals Supplier No. NBP305941PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GGA3 Polyclonal antibody specifically detects GGA3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen GGA3
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias ADP-ribosylation factor-binding protein GGA3, golgi associated, gamma adaptin ear containing, ARF binding protein 3, golgi-associated, gamma adaptin ear containing, ARF binding protein 3, KIAA0154Golgi-localized, gamma ear-containing, ARF-binding protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GGA3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 23163
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.