Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGT1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB053835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053835 Supplier Novus Biologicals Supplier No. NBP298438AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GGT1 Polyclonal specifically detects GGT1 in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GGT1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD224, CD224 antigen, D22S672, D22S732, EC 2.3.2.2, gamma-glutamyl transpeptidase, gamma-glutamyltransferase 1MGC96904, gamma-glutamyltranspeptidase 1, GGT 1, GGTMGC96963, glutamyl transpeptidase, GTG, MGC96892
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GGT1 (P07314; Pro27-Tyr568).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2678
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.