Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHDC Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB185054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185054 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185054 Supplier Novus Biologicals Supplier No. NBP282091C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GHDC Polyclonal antibody specifically detects GHDC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen GHDC
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias D11LGP1E, GH3 domain containing, GH3 domain-containing protein, homolog of mouse LGP1, LGP1D11LGP1
Host Species Rabbit
Immunogen Rabbit polyclonal GHDC antibody was raised against a 19 amino acid peptide near the amino terminus of human GHDC. The immunogen is located within amino acids 100 - 150 of GHDC. Amino Acid Squence: CEDSGEQPLPPTSNQDLGEA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84514
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.