Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHITM Antibody (42), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB388748 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388748 Supplier Novus Biologicals Supplier No. NBP312773DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GHITM Monoclonal antibody specifically detects GHITM in Invertebrate samples. It is validated for ELISA

Specifications

Antigen GHITM
Applications ELISA
Classification Monoclonal
Clone 42
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides pteronyssinus GHITM (Uniprot#: P49278; Gln19-Asp146).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27069
Target Species Invertebrate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.