Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ghrelin/Obestatin Antibody (45), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358779 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358779 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358779 Supplier Novus Biologicals Supplier No. NBP147183MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ghrelin/Obestatin Monoclonal antibody specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Ghrelin/Obestatin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 45
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46
Host Species Mouse
Immunogen Synthetic human Ser3-octanoyl ghrelin-28
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51738
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.