Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ghrelin/Obestatin Antibody, Novus Biologicals™
SDP

Catalog No. NB600813 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600813 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600813 Supplier Novus Biologicals Supplier No. NB600813
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Ghrelin/Obestatin Polyclonal specifically detects Ghrelin/Obestatin in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Ghrelin/Obestatin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2 - 5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml, Peptide ELISA 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_057446.1
Gene Alias appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46
Gene Symbols GHRL
Host Species Goat
Immunogen Peptide with sequence C-QDILWEEAKEAPADK corresponding to C-Terminus according to NP_057446.1, NP_001128413.1, NP_001128416.1, NP_001128417.1, NP_001128418.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51738
Test Specificity This antibody is expected to recognize all reported isoforms.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.