Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ghrelin Receptor/GHSR Antibody (502430), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB372985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372985 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372985 Supplier Novus Biologicals Supplier No. FAB8370L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ghrelin Receptor/GHSR Monoclonal specifically detects Ghrelin Receptor/GHSR in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Ghrelin Receptor/GHSR
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 502430
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias GH-releasing peptide receptor, Ghrelin receptor, GHRP, GHS-R, growth hormone secretagogue receptor, growth hormone secretagogue receptor type 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GHSR, Accession # NP_940799
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2693
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.