Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GHRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | GHRL |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8641
![]()
|
Proteintech Group Inc
133091AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Ghrelin is a potent stimulator of growth hormone production from the anterior pituitary gland, and is known to play a role in a number of different physiological processes. It is a 28-amino acid peptide existing in two major forms. Ghrelin plays a significant role in neurotrophy, and is essential for cognitive adaptation to changing environments and the process of learning. Ghrelin plays a significant role in various kinase dependent pathways such as AKT in activating the endothelial isoform of nitric oxide synthase.Specifications
| GHRL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9EQX0, Q9QYH7, Q9UBU3 | |
| 51738, 58991, 59301 | |
| GHRL Fusion Protein Ag4141 | |
| Primary | |
| -20°C | |
| GHRL |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Appetite regulating hormone, Ghrelin Obestatin, GHRL, Growth hormone secretagogue, Motilin related peptide, MTLRP, obestatin, Protein M46 | |
| GHRL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title