Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Giantin Rabbit anti-Human, Polyclonal, Proteintech
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 222701AP150UL
            Description
Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.Specifications
| Giantin | |
| Polyclonal | |
| Unconjugated | |
| GOLGB1 | |
| GCP, GCP372, giantin, GOLGB1, Golgin subfamily B member 1, GOLIM1, MacroGolgin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2804 | |
| -20°C | |
| Liquid | 
| Immunocytochemistry, Immunofluorescence | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14789 | |
| GOLGB1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction