Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Giantin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | Giantin |
|---|---|
| Concentration | 0.54 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3807
![]()
|
Proteintech Group Inc
222701AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.Specifications
| Giantin | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14789 | |
| 2804 | |
| Peptide | |
| Primary | |
| -20°C | |
| GOLGB1 |
| 0.54 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GCP, GCP372, giantin, GOLGB1, Golgin subfamily B member 1, GOLIM1, MacroGolgin | |
| GOLGB1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title