Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia Antibody (6231), Novus Biologicals™
SDP

Catalog No. NB10063281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063281 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063281 Supplier Novus Biologicals Supplier No. NB10063281

Mouse Monoclonal Antibody

Giardia lamblia Monoclonal specifically detects Giardia lamblia in Protozoa samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Giardia lamblia
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6231
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10-1:50, Immunocytochemistry/Immunofluorescence 1:10-1:50
Gene Alias G lamblia; Giardia intestinalis
Host Species Mouse
Immunogen 65kD Giardia antigen.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63281 detects Giardia lamblia, a flagellated protozoan parasite which infects the small intestines of humans and a variety of other mammalian hosts. Infection by G. lamblia results in giardiasis; a type of gastroenteritis that causes severe diarrhea and abdominal cramps. It has been suggested that chronic giardiasis can result in long-term growth retardation.
Target Species Protozoa
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.