Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia Antibody (BB1.1E5), Biotin, Novus Biologicals™
SDP

Catalog No. NB378474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378474 Supplier Novus Biologicals Supplier No. NBP308870B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Giardia lamblia Monoclonal antibody specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Giardia lamblia
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone BB1.1E5
Conjugate Biotin
Formulation PBS
Gene Alias G lamblia; Giardia intestinalis
Host Species Mouse
Immunogen Giardia lamblia
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.