Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibberellic Acid A4, 90%, Thermo Scientific Chemicals

Catalog No. aaj66065lb0
Click to view available options
Quantity:
5 mg

 

Used for its stimulating effect on cell division and cell elongation, seed germination and flowering in long day plants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for its stimulating effect on cell division and cell elongation, seed germination and flowering in long day plants
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 468-44-0
Molecular Formula C19H24O5
Molecular Weight (g/mol) 332.396
MDL Number MFCD00133363
InChI Key RSQSQJNRHICNNH-VGBRHYQBSA-N
PubChem CID 71306853
SMILES CC12C(CCC3(C1C(C45C3CCC(C4)C(=C)C5)C(=O)O)OC2=O)O

Specifications

Type Gibberellic Acid A4
Physical Form Powder
Color White
Quantity 5 mg
Flash Point >110°C (230°F)
Boiling Point 574°C
Solubility Information Soluble in methanol or ethanol; Also soluble in aqueous sodium bicarbonate or sodium acetate
Formula Weight 332.39
Percent Purity 90%
Chemical Name or Material Gibberellic Acid A4
EINECSNumber 207-406-9
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.