Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Trypsin (0.25%), phenol red

Click to view available options
Quantity:
100 mL
500 mL
Description
- Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas
- Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation
- This solution is formulated with phenol red indicator, and without EDTA
- Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance
- Prior to manufacturing the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis
- Lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency are all available
- Solution should be stored at -5°C to -20°C
Order Info
Shipping Condition: Dry Ice
Compliance
Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.
Specifications
Specifications
| Chelators | No EDTA |
| Content And Storage | Storage conditions: -5 to -20°C. Shelf life: 24 months from date of manufacture. |
| Cell Type | Mammalian Cells |
| Classification | Animal Origin |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Phenol Red |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction