Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gigaxonin Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291364 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291364 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291364 Supplier Novus Biologicals Supplier No. NBP149924PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gigaxonin Polyclonal antibody specifically detects Gigaxonin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Gigaxonin
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias FLJ38059, GAN1KLHL16Kelch-like protein 16, giant axonal neuropathy (gigaxonin), gigaxonin
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Gigaxonin using the numbering given in entry NP_071324.1 (GeneID 8139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8139
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.