Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GILT/IFI30 Antibody (030), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB070227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070227 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070227 Supplier Novus Biologicals Supplier No. NBP290134AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GILT/IFI30 Monoclonal specifically detects GILT/IFI30 in Human samples. It is validated for ELISA.

Specifications

Antigen GILT/IFI30
Applications ELISA
Classification Monoclonal
Clone 30
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI-30, interferon, gamma-inducible protein 30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GILT/IFI30 (P13284; Met1-Lys243).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10437
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.