Learn More
GIMAP2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 134451AP150UL
Description
This antibody detected one 28 kDa band in several tissues.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.Specifications
| GIMAP2 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP2 | |
| GIMAP2, GTPase IMAP family member 2, GTPase, IMAP family member 2, HIMAP2, IMAP2, Immunity associated protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26157 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UG22 | |
| GIMAP2 | |
| GIMAP2 Fusion Protein Ag4273 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.