Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen GIMAP2
Concentration 0.23 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-8750
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
134451AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

This antibody detected one 28 kDa band in several tissues.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications

Specifications

GIMAP2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q9UG22
26157
GIMAP2 Fusion Protein Ag4273
Primary
-20°C
GIMAP2
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GIMAP2, GTPase IMAP family member 2, GTPase, IMAP family member 2, HIMAP2, IMAP2, Immunity associated protein 2
GIMAP2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.