Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP4 Antibody (OTI1A11), Novus Biologicals™
SDP

Catalog No. NBP245854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245854 Supplier Novus Biologicals Supplier No. NBP245854
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence

GIMAP4 Monoclonal specifically detects GIMAP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen GIMAP4
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150, Immunofluorescence 1:100
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias GTPase IMAP family member 4, GTPase, IMAP family member 4, HIMAP4, human immune associated nucleotide 1, IAN-1, IAN1FLJ11110, IMAP4hIAN1, immunity associated protein 4, Immunity-associated nucleotide 1 protein, Immunity-associated protein 4
Gene Symbols GIMAP4
Host Species Mouse
Immunogen Full length human recombinant protein of human GIMAP4(NP_060796) produced in HEK293T cell.
Molecular Weight of Antigen 37.4 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55303
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.