Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP4 Antibody (OTI1C6), PerCP, Novus Biologicals™
SDP

Catalog No. NB271997PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271997PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271997PCP Supplier Novus Biologicals Supplier No. NBP271997PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIMAP4 Monoclonal specifically detects GIMAP4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GIMAP4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C6
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias GTPase IMAP family member 4, GTPase, IMAP family member 4, HIMAP4, human immune associated nucleotide 1, IAN-1, IAN1FLJ11110, IMAP4hIAN1, immunity associated protein 4, Immunity-associated nucleotide 1 protein, Immunity-associated protein 4
Gene Symbols GIMAP4
Host Species Mouse
Immunogen Full length recombinant protein of human GIMAP4 (NP_060796) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55303.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.