Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, Giordano, pH 6.4 (M/15), for Wright Staining, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Buffer |
CAS | 7732-18-5, 7778-77-0, 7558-79-4 |
For Use With (Application) | Wright Staining |
Physical Form | Solution |
Quantity | 1 L |
Packaging | Natural Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction