Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIP Antibody (14), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB154843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154843 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154843 Supplier Novus Biologicals Supplier No. NBP312092G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIP Monoclonal antibody specifically detects GIP in Human samples. It is validated for ELISA

Specifications

Antigen GIP
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Host Species Mouse
Immunogen Synthetic human gastric inhibitory polypeptide
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.