Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIP Antibody (4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP221617AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221617AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221617AG Supplier Novus Biologicals Supplier No. NBP221617AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIP Monoclonal specifically detects GIP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GIP
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4
Conjugate Alexa Fluor 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Gene Symbols GIP
Host Species Mouse
Immunogen Synthetic human gastric inhibitory polypeptide adsorbed onto aluminum hydroxide gel.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Test Specificity Cross-reacts up to 10% with PACAP and human GLP-2 but <0.1% with glucagon, GLP-1 or VIP.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.