Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIP Antibody (4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019252 0.025 mg
NBP221617 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019252 Supplier Novus Biologicals Supplier No. NBP2216170.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIP Monoclonal specifically detects GIP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GIP
Applications ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone 4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.14M NaCl with 0.05% Sodium Azide
Gene Alias gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Gene Symbols GIP
Host Species Mouse
Immunogen Synthetic human gastric inhibitory polypeptide adsorbed onto aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Test Specificity Cross-reacts up to 10% with PACAP and human GLP-2 but <0.1% with glucagon, GLP-1 or VIP.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.