Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIP Antibody, Novus Biologicals™
SDP

Catalog No. NB036597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036597 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036597 Supplier Novus Biologicals Supplier No. NBP289006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIP Polyclonal specifically detects GIP in Mammal samples. It is validated for Immunohistochemistry.

Specifications

Antigen GIP
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat serum
Gene Alias gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Host Species Rabbit
Immunogen Natural pig GIP.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Target Species Mammalia
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.