Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001353 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001353 Supplier Novus Biologicals Supplier No. NB1001353
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

GIPC1 Polyclonal specifically detects GIPC1 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen GIPC1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.1 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 10 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O14908
Gene Alias C19orf3chromosome 19 open reading frame 3, GAIP C-terminus-interacting protein, GIPC PDZ domain containing family, member 1, GIPCRGS-GAIP-interacting protein, GLUT1 C-terminal binding protein, GLUT1CBP, IGF-1 receptor interacting protein 1, MGC15889, MGC3774, NIP, PDZ domain-containing protein GIPC1, regulator of G-protein signalling 19 interacting protein 1, RGS19IP1IIP-1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP-2RGS19-interacting protein 1
Gene Symbols GIPC1
Host Species Goat
Immunogen Peptide with sequence C-GAIGDAKVGRY corresponding to C-Terminus according to NP_974197.1, NP_974199.1, NP_974196.1, NP_974198.1, NP_974223.1.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10755
Test Specificity This antibody is expected to recognize both isoform 1 (NP_005707.1, NP_974197.1 and NP_974199.1) and isoform 2 (NP_974196.1, NP_974198.1 and NP_974223.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.