Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC2 Antibody (1G10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2222A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2222A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2222A30 Supplier Novus Biologicals Supplier No. NBP222552AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIPC2 Monoclonal specifically detects GIPC2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen GIPC2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1G10
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ20075, GIPC PDZ domain containing family, member 2, PDZ domain protein GIPC2, PDZ domain-containing protein GIPC2, semaF cytoplasmic domain associated protein 2, semaphorin cytoplasmic domain associated protein 2, SEMCAP2, SEMCAP-2
Gene Symbols GIPC2
Host Species Mouse
Immunogen Recombinant human GIPC2 (1-315aa) purified from E. coli
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54810
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.