Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Antibody (591827), Alexa Fluor™ 750, R&D Systems™
SDP

Catalog No. MB82102A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB82102A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB82102A750 Supplier Novus Biologicals Supplier No. MAB82102AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GIPR
Applications Flow Cytometry
Classification Monoclonal
Clone 591827
Conjugate Alexa Fluor 750
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Gene Symbols GIPR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2696
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.