Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Antibody (591853), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB373378 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373378 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373378 Supplier Novus Biologicals Supplier No. FAB8210Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen GIPR
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 591853
Conjugate DyLight 680
Dilution Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GIPR., Met1-Cys466, Accession # P48546
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2696
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.