Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Antibody (Gipg013), Biotin, Novus Biologicals™
SDP

Catalog No. NB015300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015300 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015300 Supplier Novus Biologicals Supplier No. NBP281068B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GIPR
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone Gipg013
Conjugate Biotin
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Gene Symbols GIPR
Host Species Rabbit
Immunogen Human GIPR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2696
Test Specificity The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.