Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Girard's Reagent T, 99%

Catalog No. AC119872500
Click to view available options
Quantity:
50 g
100 g
250 g

Catalog No. AC119872500

Supplier: Thermo Scientific Chemicals 119872500

Only null left
Add to Cart

CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-46-6
Molecular Formula C5H14ClN3O
Molecular Weight (g/mol) 167.64
MDL Number MFCD00012009
InChI Key YSULOORXQBDPCU-UHFFFAOYSA-N
Synonym girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide
PubChem CID 67156
SMILES [Cl-].C[N+](C)(C)CC(=O)NN

Specifications

Assay Percent Range 99%
Linear Formula H2NNHCOCH2N(CH3)3Cl
Fieser 01,410
Merck Index 15, 4465
IUPAC Name (2-hydrazinyl-2-oxoethyl)-trimethylazanium;chloride
Formula Weight 167.64
Packaging Glass bottle
Quantity 250 g
Chemical Name or Material Girard's Reagent T
Hazard Category Warning
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.