Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Gisadenafil besylate

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent PDE5 inhibitor (IC50 = 1.23 nM). Shows >100-fold selectivity for PDE5 over PDE6. Orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 677.79 |
Purity | >99% |
Molecular Weight (g/mol) | 677.79 |
Chemical Name or Material | Gisadenafil besylate |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction