Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GIT 27
GREENER_CHOICE

Catalog No. 327010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-701-0 10 mg
32-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-701-0 Supplier Tocris Bioscience™ Supplier No. 3270/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Immunomodulator; reduces production of pro-inflammatory cytokines

Orally active immunomodulatory agent that primarily targets macrophages. Inhibits TNF-α secretion via interference of macrophage toll-like receptor (TLR) 4 and TLR 2/6 signaling pathway. Also reduces the secretion of pro-inflammatory cytokines IL1-β, IL-10 and IFN-γ. Antidiabetogenic; prevents IL-β and IFN-γ-induced pancreatic islet cell death in vitro.

Chemical Identifiers

CAS 6501-72-0
Molecular Formula C11H11NO3

Specifications

Quantity 10 mg
Formula Weight 205.21
Purity >99%
Molecular Weight (g/mol) 205.21
Chemical Name or Material GIT 27
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.