Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT1 Antibody (924640), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB302551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302551 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302551 Supplier Novus Biologicals Supplier No. FAB8508G

Mouse Monoclonal Antibody

GIT1 Monoclonal antibody specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen GIT1
Applications Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 924640
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28964
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.