Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP119143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119143 Supplier Novus Biologicals Supplier No. NBP119143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIT2 Polyclonal specifically detects GIT2 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).

Specifications

Antigen GIT2
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_476510.1
Gene Alias ARF GAP GIT2, ARF GTPase-activating protein GIT2, CAT2, CAT-2, cool-associated, tyrosine phosphorylated protein 2, Cool-interacting tyrosine-phosphorylated protein 2, DKFZp686G01261, G protein-coupled receptor kinase interacting ArfGAP 2, G protein-coupled receptor kinase interactor 2, G protein-coupled receptor kinase-interactor 2, KIAA0148GRK-interacting protein 2, MGC760
Gene Symbols GIT2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human G protein-coupled receptor kinase interacting ArfGAP 2 using the numbering given in entry NP_476510.1 (GeneID 9815).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9815
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Chinese hamster, and Naked mole rat.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.