Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR Ligand/TNFSF18 Antibody (994572), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313701 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313701 Supplier Novus Biologicals Supplier No. FAB21774PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry
Classification Monoclonal
Clone 994572
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8995
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.