Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR Ligand/TNFSF18 Antibody (MM0312-4Q25), HRP, Novus Biologicals™
SDP

Catalog No. NB361524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361524 Supplier Novus Biologicals Supplier No. NBP211965H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry, CyTOF, Neutralization

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry, CyTOF, Neutralization
Classification Monoclonal
Clone MM0312-4Q25
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready, Block/Neutralize
Formulation PBS
Gene Alias Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Host Species Mouse
Immunogen Human recombinant GITR Ligand/TNFSF18 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8995
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.