Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR/TNFRSF18 Antibody (002), Alexa Fluor™ 700
SDP

Catalog No. NB071308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB071308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB071308 Supplier Novus Biologicals Supplier No. NBP290234AF700

Rabbit Monoclonal Antibody

GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for ELISA.

Specifications

Antigen GITR/TNFRSF18
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GITR/TNFRSF18 (NP_683700.1; Met1-Glu161).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.