Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR/TNFRSF18 Antibody (108619), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB174011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174011 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174011 Supplier Novus Biologicals Supplier No. FAB5241Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen GITR/TNFRSF18
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 108619
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GITR/TNFRSF18, Met1-His153, Accession # O35714
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.