Learn More
Description
Specifications
Specifications
| Antigen | GITR/TNFRSF18 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DTA-1 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 |
| Gene Symbols | TNFRSF18 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
