Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR/TNFRSF18 Antibody (DTA-1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP226661IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226661IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226661IR Supplier Novus Biologicals Supplier No. NBP226661IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen GITR/TNFRSF18
Applications Flow Cytometry
Classification Monoclonal
Clone DTA-1
Conjugate DyLight 755
Dilution Flow Cytometry
Gene Accession No. Q9Y5U5
Gene Alias Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Gene Symbols TNFRSF18
Host Species Rat
Immunogen Mouse CD25+ CD4+ T cell line NP_683700
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.